Learn More
Aviva Systems Biology Corp EEA1 Antibody - N-terminal region (ARP58126_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58126_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EEA1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human EEA1. Alternate names: ZFYVE2, MST105, MSTP105ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-286-440