Learn More
Aviva Systems Biology Corp EEA1 Antibody - middle region (ARP58125_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58125_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EEA1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human EEA1. Alternate names: ZFYVE2, MST105, MSTP105ApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Rabbit, Rat
Catalog No. 50-014-62243