Learn More
Aviva Systems Biology Corp EED Antibody - N-terminal region (ARP38384_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38384_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EED. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human EED. Alternate names: HEED, WAIT1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56632