Learn More
Aviva Systems Biology Corp Eed Antibody - N-terminal region (ARP35696_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35696_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Eed. It was validated on Western Blot. . Alternate names: ENSMUSG00000039373, l(7)5Rn, l7Rn5, luskApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-469