Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC EED226-COOH | 2467965-71-3 | 99.9% | 100 MG
SDP

Catalog No. 5000341126
Encompass_Preferred

EED226-COOH is a ligand derived from EED226 for the target protein EED. It functions in PROTAC by binding to a ligand for VHL via a linker, forming UNC6852 to degrade PRC2. This product is for research use only.

  • Molecular weight: 335.32
  • Formula: C17H13N5O3
  • Appearance: Solid
  • Color: Off-white to light yellow
  • Solubility in DMSO: 125 mg/mL (372.78 mM)
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 2 years, -20°C for 1 year
  • Target: EED

Catalog No. 50-003-41126 Supplier Medchemexpress LLC Supplier No. HY130979100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

EED226-COOH is a ligand derived from EED226 for the target protein EED. It functions in PROTAC by binding to a ligand for VHL via a linker, forming UNC6852 to degrade PRC2. This product is for research use only.

  • Molecular weight: 335.32
  • Formula: C17H13N5O3
  • Appearance: Solid
  • Color: Off-white to light yellow
  • Solubility in DMSO: 125 mg/mL (372.78 mM)
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 2 years, -20°C for 1 year
  • Target: EED

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.