Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC EED226-COOH | 2467965-71-3 | 99.9% | 50 MG
SDP

Catalog No. 5000338258
Encompass_Preferred

EED226-COOH is a ligand derived from EED226, designed for target protein EED in PROTAC applications. It forms UNC6852 by binding to a VHL ligand via a linker, facilitating the degradation of PRC2. This product is intended for research use only and is not for patient administration.

  • Derived ligand for target protein EED
  • Utilized in PROTAC (proteolysis-targeting chimera) research
  • Forms UNC6852 through binding with a VHL ligand
  • Facilitates degradation of PRC2
  • Available as an off-white to light yellow solid
  • Molecular weight is 335.32 g/mol
  • Chemical formula is C17H13N5O3
  • Soluble in DMSO at 125 mg/mL

Catalog No. 50-003-38258 Supplier Medchemexpress LLC Supplier No. HY13097950MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

EED226-COOH is a ligand derived from EED226, designed for target protein EED in PROTAC applications. It forms UNC6852 by binding to a VHL ligand via a linker, facilitating the degradation of PRC2. This product is intended for research use only and is not for patient administration.

  • Derived ligand for target protein EED
  • Utilized in PROTAC (proteolysis-targeting chimera) research
  • Forms UNC6852 through binding with a VHL ligand
  • Facilitates degradation of PRC2
  • Available as an off-white to light yellow solid
  • Molecular weight is 335.32 g/mol
  • Chemical formula is C17H13N5O3
  • Soluble in DMSO at 125 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.