Learn More
Bachem EEDQ
SDP
Supplier:  Bachem 4015690.0500
Encompass_Preferred
EEDQ, N-Ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, 16357-59-8, C14H17NO3, Mr 247.29, 500g. EEDQ permits the coupling of acylamino acids with amino acid esters in high yield without concomitant racemization. In a study aiming at the optimization of the coupling of 3,5-dinitrobenzoyl-Leu to 3-aminopropyl-silica (Yang et al.) significant levels of racemization were observed with all the tested coupling reagents except for EEDQ .
Catalog No. 50-194-7381