Learn More
Aviva Systems Biology Corp EEF1A1 Antibody - C-terminal region (ARP48166_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48166_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against EEF1A1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human EEF1A1. Alternate names: CCS-3, CCS3, EEF-1, EEF1A, EF-Tu, EF1A, FLJ25721, GRAF-1EF, HNGC:16303, LENG7, MGC102687, MGC131894, MGC16224, PTI1, eEF1A-1, EE1A1ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-59738