Learn More
Aviva Systems Biology Corp EEF1A2 Antibody - middle region (ARP48202_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48202_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EEF1A2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human EEF1A2. Alternate names: EEF1AL, EF-1-alpha-2, EF1A, HS1, STN, STNLApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59754