Learn More
Aviva Systems Biology Corp EEF1B2 Antibody - middle region (ARP52399_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52399_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EEF1B2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human EEF1B2. Alternate names: EEF1B, EEF1B1, EF1BApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit
Catalog No. 50-014-60726