Learn More
Aviva Systems Biology Corp EEF1D Antibody - C-terminal region (ARP62273_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62273_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EEF1D. It was validated on Western Blot. . Alternate names: EF-1D, EF1D, FLJ20897, FP1047ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Yeast
Catalog No. 50-287-642