Learn More
Aviva Systems Biology Corp EEF1E1 Antibody - N-terminal region (ARP62511_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62511_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EEF1E1. It was validated on Western Blot. . Alternate names: AIMP3, P18ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-701