Learn More
Aviva Systems Biology Corp EEF1G Antibody - N-terminal region (ARP48167_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48167_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against EEF1G. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human EEF1G. Alternate names: EF1G, GIG35ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59739