Learn More
Aviva Systems Biology Corp EEF2 Antibody - N-terminal region (ARP58457_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58457_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EEF2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human EEF2. Alternate names: EEF-2, EF2, EF-2ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-53769