Learn More
Aviva Systems Biology Corp EEFSEC Antibody - C-terminal region (ARP65659_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP65659_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EEFSEC. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C-terminal region of Human EEFSEC. Alternate names: EFSEC, SELBApplicationWB. Species reactivity: Human
Catalog No. 50-288-531