Learn More
Aviva Systems Biology Corp Eepd1 Antibody - N-terminal region (ARP48411_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48411_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Eepd1. It was validated on Western Blot. . Alternate names: 2310005P05Rik, AI481005ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-359