Learn More
Selleck Chemical LLC Eeyarestatin I-E1389-5MG
SDP
Catalog No. 503134442
Encompass_Preferred
Eeyarestatin I is a potent inhibitor of endoplasmic reticulum-associated protein degradation (ERAD) Eeyarestatin I also inhibits Sec61 translocon
Catalog No. 50-313-4442 Supplier Selleck Chemical LLC Supplier No. E13895MG
Eeyarestatin I is a potent inhibitor of endoplasmic reticulum-associated protein degradation (ERAD) Eeyarestatin I also inhibits Sec61 translocon