Learn More
AnaSpec EndoClear™ Human Recombinant Alpha-Synuclein Protein, High Purity, Low Endotoxin Levels, Unit Size: 1000 µg
Supplier:  AnaSpec AS555551000
Encompass
The EndoClear™ Human Recombinant alpha-Synuclein Protein is a high-purity, recombinant protein crucial for research on neurodegenerative diseases like Parkinson's. Supplied as a lyophilized powder, it supports scientific investigations into fibrillation, oligomerization, and cell toxicity, featuring low endotoxin levels that ensure suitability for sensitive applications. This product stands out due to its additional Glycine residue at the N-terminus, enhancing its functional profile for researchers.
Catalog No. NC1157420