Learn More
BIOHIPPO EF1 -Luc-RFP-BSD
SDP
Catalog No. 5000331008
Encompass
EF1 -Luc-RFP-BSD is a fluorescence and luciferase dual imaging reporter lentivirus based on the LFV-V1a vector layout under control of the EF1 promoter containing a Luc-T2A-RFP-P2A-BSD expression cassette and Blasticidin resistance marker This construct co expresses luciferase and fluorescent reporter proteins to enable correlated bioluminescent and fluorescent readouts in the same cell population Each vial provides a nominal titer of 2x10E6 transducing units TU The lentiviral particles are supplied ready to use for stable gene delivery to mammalian cells and can facilitate generation of long term reporter cell lines They are well suited for in vitro cell based assays in vivo optical imaging high content screening and mechanism of action studies This product is intended for research use only and is not for diagnostic or therapeutic procedures
Catalog No. 50-003-31008 Supplier BIOHIPPO Supplier No. BHV1940018110
EF1 -Luc-RFP-BSD is a fluorescence and luciferase dual imaging reporter lentivirus based on the LFV-V1a vector layout under control of the EF1 promoter containing a Luc-T2A-RFP-P2A-BSD expression cassette and Blasticidin resistance marker This construct co expresses luciferase and fluorescent reporter proteins to enable correlated bioluminescent and fluorescent readouts in the same cell population Each vial provides a nominal titer of 2x10E6 transducing units TU The lentiviral particles are supplied ready to use for stable gene delivery to mammalian cells and can facilitate generation of long term reporter cell lines They are well suited for in vitro cell based assays in vivo optical imaging high content screening and mechanism of action studies This product is intended for research use only and is not for diagnostic or therapeutic procedures