Learn More
BIOHIPPO EF1a-KRAS-V5/RFP
SDP
Catalog No. 5000331714
Encompass
EF1a-KRAS-V5/RFP lentiviral vector expressing V5 under EF1a promoter Each vial provides a nominal titer of 2x10E6 transducing units TU Human KRAS ORF cDNA lentivirus (wild type) Lentiviral particles packaged with expression vector for wild type human KRAS proto-oncogene GTPase (KRAS) transcript variant a An EF1 promoter drives expression of KRAS fused to a C-terminal V5 tag and a GFP reporter which is separated from the ORF cDNA by a self-cleaving peptide These lentiviral particles allow for easy transduction and high stable expression of the indicated ORF cDNA in mammalian cells The lentiviral vectors have been optimized to provide high gene expression and reliable genome integration allowing for easy establishment of long-term stable cell lines with consistent read out of fluorescent (GFP RFP) or luminescent (firefly luc) reporters
Catalog No. 50-003-31714 Supplier BIOHIPPO Supplier No. BHV194001002
EF1a-KRAS-V5/RFP lentiviral vector expressing V5 under EF1a promoter Each vial provides a nominal titer of 2x10E6 transducing units TU Human KRAS ORF cDNA lentivirus (wild type) Lentiviral particles packaged with expression vector for wild type human KRAS proto-oncogene GTPase (KRAS) transcript variant a An EF1 promoter drives expression of KRAS fused to a C-terminal V5 tag and a GFP reporter which is separated from the ORF cDNA by a self-cleaving peptide These lentiviral particles allow for easy transduction and high stable expression of the indicated ORF cDNA in mammalian cells The lentiviral vectors have been optimized to provide high gene expression and reliable genome integration allowing for easy establishment of long-term stable cell lines with consistent read out of fluorescent (GFP RFP) or luminescent (firefly luc) reporters