Learn More
Gentarget EF1A-LUCIFERASE -2A-GFP 200UL
Catalog No. NC0503250
Encompass
Pre-made lentiviral particles bicistronically expressing firefly luciferase and GFP fluorescent protein under EF1a promoter, with Puromycin selection marker under Rsv promoter, provided in PBS solution (200ul).
Catalog No. NC0503250 Supplier Gentarget Supplier No. LVP437PBS
Pre-made lentiviral particles bicistronically expressing firefly luciferase and GFP fluorescent protein under EF1a promoter, with Puromycin selection marker under Rsv promoter, provided in PBS solution (200ul).