Learn More
Amsbio LLC EF1a-Luciferase (firefly)-2A-RFP (Neo), Concentrated Lentivirus, 1x10^8 IFU/ml x 200ul
Catalog No. NC3913730
Encompass
Pre-made Lentivirus bicistronically expressing firefly luciferase and RFP fluorescent protein under EF1a promoter, with Neomycin selection marker under Rsv promoter, provided in PBS solution.
Catalog No. NC3913730 Supplier Amsbio LLC Supplier No. LVP441PBS
Pre-made Lentivirus bicistronically expressing firefly luciferase and RFP fluorescent protein under EF1a promoter, with Neomycin selection marker under Rsv promoter, provided in PBS solution.