Learn More
TARGETMOL CHEMICALS INC EF24 10MG
SDP
Catalog No. 502577054
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. EF24 (35-Bis[(2-fluorophenyl)Methylene]-4-piperidinone) treatment increases the levels of activated caspase 3 and 9 and decreases the phosphorylated forms of MEK1 and ERK. EF24 shows potent anti-tumor activity in oral squamous cell carcinoma (OSCC) via deactivation of the MAPK/ERK signaling pathway. purity: 98%
Catalog No. 50-257-7054 Supplier TARGETMOL CHEMICALS INC Supplier No. T2724210MG
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. EF24 (35-Bis[(2-fluorophenyl)Methylene]-4-piperidinone) treatment increases the levels of activated caspase 3 and 9 and decreases the phosphorylated forms of MEK1 and ERK. EF24 shows potent anti-tumor activity in oral squamous cell carcinoma (OSCC) via deactivation of the MAPK/ERK signaling pathway. purity: 98%