Learn More
Medchemexpress LLC Efaroxan hydrochloride | 89197-00-2 | 99.8% | 1 ML
SDP
Catalog No. 5000413002
Encompass_Preferred
Efaroxan hydrochloride is a potent, selective, and orally active α2-adrenoceptor antagonist with antidiabetic activity. It is also a selective I1-Imidazoline receptor antagonist. This product can be utilized for research related to cardiovascular disease.
  • Potent, selective, and orally active α2-adrenoceptor antagonist
  • Demonstrates antidiabetic activity
  • Selective I1-Imidazoline receptor antagonist
  • Suitable for research related to cardiovascular disease
Catalog No. 50-004-13002 Supplier Medchemexpress LLC Supplier No. HYB1416A10MM/1ML
Efaroxan hydrochloride is a potent, selective, and orally active α2-adrenoceptor antagonist with antidiabetic activity. It is also a selective I1-Imidazoline receptor antagonist. This product can be utilized for research related to cardiovascular disease.
  • Potent, selective, and orally active α2-adrenoceptor antagonist
  • Demonstrates antidiabetic activity
  • Selective I1-Imidazoline receptor antagonist
  • Suitable for research related to cardiovascular disease