Learn More
Medchemexpress LLC Efaroxan hydrochloride | 89197-00-2 | 99.80% | 25 MG
SDP
Catalog No. 5000413831
Encompass_Preferred
Efaroxan hydrochloride is a potent, selective, and orally active α2-adrenoceptor antagonist, demonstrating antidiabetic activity. It also acts as a selective I1-Imidazoline receptor antagonist. This compound can be utilized in research related to cardiovascular disease.
  • Potent, selective, and orally active α2-adrenoceptor antagonist.
  • Possesses antidiabetic activity.
  • Selective I1-Imidazoline receptor antagonist.
  • Applicable for research in cardiovascular disease.
  • Binds to I1-imidazoline and α2-adrenergic receptors with Kᵢ values of 0.15 nM and 5.6 nM, respectively, in bovine rostral ventrolateral medulla membranes.
  • Increases plasma insulin levels in conscious fed and fasted rats without significantly affecting plasma glucose levels.
Catalog No. 50-004-13831 Supplier Medchemexpress LLC Supplier No. 25MGHYB1416A
Efaroxan hydrochloride is a potent, selective, and orally active α2-adrenoceptor antagonist, demonstrating antidiabetic activity. It also acts as a selective I1-Imidazoline receptor antagonist. This compound can be utilized in research related to cardiovascular disease.
  • Potent, selective, and orally active α2-adrenoceptor antagonist.
  • Possesses antidiabetic activity.
  • Selective I1-Imidazoline receptor antagonist.
  • Applicable for research in cardiovascular disease.
  • Binds to I1-imidazoline and α2-adrenergic receptors with Kᵢ values of 0.15 nM and 5.6 nM, respectively, in bovine rostral ventrolateral medulla membranes.
  • Increases plasma insulin levels in conscious fed and fasted rats without significantly affecting plasma glucose levels.