Learn More
Selleck Chemical LLC Efavirenz 100mg 154598-52-4 Sustiva, Stocrin, DMP-266, DMP 266
SDP
Catalog No. 501364172
Encompass_Preferred
Efavirenz (Sustiva, Stocrin, DMP-266) is a synthetic non-nucleoside reverse transcriptase (RT) inhibitor with antiviral activity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4172 Supplier Selleck Chemical LLC Supplier No. S4685100MG
Efavirenz (Sustiva, Stocrin, DMP-266) is a synthetic non-nucleoside reverse transcriptase (RT) inhibitor with antiviral activity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.