Learn More
Apexbio Technology LLC Efavirenz, 10mg. Cas: 154598-52-4 MFCD: MFCD05662344. Reverse transcriptase inhibitor
SDP
Supplier:  Apexbio Technology LLC B111910
Encompass_Preferred
Efavirenz is a highly potent inhibitor of human immunodeficiency virus type 1 reverse transcriptase with Ki value of 2.93nM [1].Efavirenz is an antiretroviral drug approved by the FDA in 1998. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5206