Learn More
AdipoGen Efavirenz
SDP
Catalog No. 502562184
Encompass_Preferred
Chemical. CAS 154598-52-4. Formula C14H9ClF3NO2. MW 315.7. Efavirenz is a non-nucleoside reverse transcriptase inhibitor NNRTI that exhibits antiviral and anticancer activities. Efavirenz binds to wild-type and mutant HIV-1 RTs Ki= 2.93nM and 3.85-56.5nM, respectively. It inhibits wild-type and mutant HIV-1 viral replication in MT-4 human T lymphoid cells IC95s = 1.5-1,500 nM. Efavirenz also prevents RNA plus-strand initiation with an IC50 value of 17nM. Efavirenz is an anti-HIV drug, commonly used in combination therapy for AIDS treatment. It is part of highly active antiretroviral therapy HAART for the treatment of a human immunodeficiency virus HIV type 1. In various cancer cell lines, Efavirenz inhibits cellular proliferation and increases activation of p53. CARD8 inflammasome senses HIV-1 protease activity. In HIV1-infected cells, CARD8 cannot detect the virus because the viral protease remains inactive as a subunit of unprocessed Gag-Pol polyprotein.
Catalog No. 50-256-2184 Supplier AdipoGen Supplier No. AGCR13751M050
Chemical. CAS 154598-52-4. Formula C14H9ClF3NO2. MW 315.7. Efavirenz is a non-nucleoside reverse transcriptase inhibitor NNRTI that exhibits antiviral and anticancer activities. Efavirenz binds to wild-type and mutant HIV-1 RTs Ki= 2.93nM and 3.85-56.5nM, respectively. It inhibits wild-type and mutant HIV-1 viral replication in MT-4 human T lymphoid cells IC95s = 1.5-1,500 nM. Efavirenz also prevents RNA plus-strand initiation with an IC50 value of 17nM. Efavirenz is an anti-HIV drug, commonly used in combination therapy for AIDS treatment. It is part of highly active antiretroviral therapy HAART for the treatment of a human immunodeficiency virus HIV type 1. In various cancer cell lines, Efavirenz inhibits cellular proliferation and increases activation of p53. CARD8 inflammasome senses HIV-1 protease activity. In HIV1-infected cells, CARD8 cannot detect the virus because the viral protease remains inactive as a subunit of unprocessed Gag-Pol polyprotein.