Learn More
Medchemexpress LLC HY-10572 5mg Medchemexpress, Efavirenz CAS:154598-52-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY105725MG
Encompass_Preferred
Medchemexpress, HY-10572 5mg Efavirenz CAS:154598-52-4 Efavirenz is a potent inhibitor of the wild-type HIV-1 reverse transcriptase with a Ki of 2.93 nM and exhibits an IC95 of 1.5 nM for the inhibition of HIV-1 replicative spread in cell culture. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8291