Learn More
Aviva Systems Biology Corp EFCAB3 Antibody - N-terminal region (ARP53251_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53251_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EFCAB3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human EFCAB3. Alternate names: FLJ25818, MGC126801, MGC126827ApplicationWB. Species reactivity: Human, Rat
Catalog No. 50-285-815