Learn More
Aviva Systems Biology Corp CRACR2A Antibody - middle region (ARP48422_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48422_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EFCAB4B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human EFCAB4B. Alternate names: FLJ33805, MGC4266, CRACR2AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59819