Learn More
Aviva Systems Biology Corp EFEMP1 Antibody - middle region (ARP41341_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41341_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EFEMP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human EFEMP1. Alternate names: DHRD, DRAD, FBLN3, FBNL, FLJ35535, MGC111353, MLVT, MTLV, S1-5ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-57811