Learn More
Aviva Systems Biology Corp EFEMP2 Antibody - middle region (ARP55351_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55351_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EFEMP2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human EFEMP2. Alternate names: FBLN4, MBP1, UPH1, ARCL1BApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61474