Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Effects of Alcohol on C. elegans Experiment Kit
SDPSCI_ED

Catalog No. S29024 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
10 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S29024 10 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S29024 Supplier Edvotek™ Supplier No. 851
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Requires

Covered box, microscopes, timers, 70% ethanol, waterbath, and incubator (optional).

The objective of this experiment is to observe and record the effects of alcohol on normal and alcohol mutant strains of Caenorhabditis elegans. Edvotek™ Effects of Alcohol on C. elegans Experiment Kit contains LIVE materials and must be requested 1 week prior to lab.

  • For 10 Lab Groups
  • Growing Bacteria: overnight; Plating Worms: 15 min.; Worm Growth: 3-4 Days; Alcohol Experiment: 45-60 min.
  • Level: Intermediate

Recommended Storage

4˚C. 3 months. (Some Components)

Specifications

Grade 9 to Undergraduate
Includes Instructions, C. elegans-normal, C. elegans Alcohol-resistant, E.coli OP50 BactoBeads™, S-Buffer, 10% Alcohol, ReadyPour NGM Agar, NGM Medium Salts, Petri Dishes, Large/Small Transfer Pipettes, 1.5mL Microcentrifuge Tubes, Cell Counting Chamber, Sterile Loops, 15mL centrifuge tubes, and 10mL pipette.
Time Range Growing Bacteria: overnight; Plating Worms: 15 min.; Worm Growth: 3-4 Days; Alcohol Experiment: 45-60 min.
Class Size 10 Lab Groups
Product Type Effects of alcohol on C. elegans Experiment Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.