Learn More
Anton Paar Efficiency meets compliance: Anton Paar Spectroscopy Suite
Catalog No. 5000580524
Encompass_Preferred
AP Spectroscopy Suite: Lyza 7000/3000 and Cora 5001Seamless Integration into your IT infrastructureCompliance with critical standards for seamless regulatory adherenceCompliance with 21 CFR Part 11 and EU GMP Vol. 4 Annex 11Full traceability (audit trail, library and method versioning, etc.)Permission management via active directoryData integrity: All data stored in a secure SQL databaseFully compatible with AP Connect
Catalog No. 50-005-80524 Supplier Anton Paar Supplier No. 262502
AP Spectroscopy Suite: Lyza 7000/3000 and Cora 5001Seamless Integration into your IT infrastructureCompliance with critical standards for seamless regulatory adherenceCompliance with 21 CFR Part 11 and EU GMP Vol. 4 Annex 11Full traceability (audit trail, library and method versioning, etc.)Permission management via active directoryData integrity: All data stored in a secure SQL databaseFully compatible with AP Connect