Learn More
Gibco™ Efficient-Pro™ AGT™ Feed 1
Description
- A next generation CHO feed used in the production of monoclonal antibodies
- Feed 1 offers significant performance and ease-of-use improvements not seen in existing Gibco™ or competitor products
- Formulated as a chemically-defined, animal-origin free cell culture feed used in the bioproduction of mAbs & other therapeutics in traditional batch & fed-batch bioprocessing workflows
- Free of growth factors or insulin to ensure batch-to-batch consistency and production efficiency
- Designed to work optimally with Efficient-Pro™ Medium, Efficient-Pro™ AGT™ Feed 1 has been formulated specifically for use in CHO-K1 cell lines
- The Efficient-Pro system increases titer and specific productivity, improves protein quality and metabolite profiles
- System is easier to use with less solubility issues than other CHO media and feed offerings
- Increase ease-of-use with Gibco’s™ trademarked, patented Advanced Granulation Technology™ (AGT™) for auto-pH and simplified dissolution
- Sustain cell viability for longer bioreactor runs & maximized titer
- Demonstrated to improve titers in a CHO-K1 cell line up to 147% when paired with Efficient-Pro™ Medium compared to an industry-leader’s media and feed system
- Improve downstream outputs such as protein glycosylation, charge-variants & aggregation
- Speed-up process development and streamline transfer to manufacturing scale
- Increase flexibility with a range of format & packaging configurations
- Decrease cell shock with simplified adaptation protocols
Specifications
Specifications
| Content And Storage | Store in the dark at 2–8°C. |
| Cell Line | CHO-DG44, CHO-S, CHO-K1, GS-CHO |
| Classification | Animal Origin-free, Chemically-defined, Protein-free, Serum-free |
| Culture Type | Mammalian Cell Culture |
| Form | AGT |
| Quantity | 100 L |
| Shipping Condition | Room temperature |
Frequently Asked Questions (FAQs)
Efficient-Pro AGT Feed 1 has been designed specifically for CHO-S and DG-44 cells; however, it has been demonstrated to improve performance in other CHO variants and even other parental cell lines.
We recommend continuing current supplementation protocols including insulin and other growth factors when using Efficient-Pro AGT Feed 1.
The shelf-life of Efficient-Pro AGT Feed 1 is 12 months from the date of manufacture. The expiration date is provided on the product label.
The comprehensive feed protocol for Efficient-Pro AGT Feed 1 is included in the user guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0025907_EfficientProAGTFeed1_UG.pdf). We recommend first designing a small-scale experiment using an Ambr15 Microbioreactor or other small vessel with concentrations between 1.5% to 3.0% of Efficient-Pro AGT Feed 1 in 0.5% increments beginning on day 3, with daily feeds through harvest. Once the specific feeding strategy is tailored to your CHO cell system for optimized results, cultures can be scaled up to larger vessels and bioreactors.
Efficient-Pro AGT Feed 1 is a chemically-defined, animal-origin-free supplement that has been formulated to work specifically with CHO-S and DG-44 cells for production of proteins. Efficient-Pro AGT Feed 1 has been designed to work optimally when paired with Efficient-Pro AGT Medium (https://www.thermofisher.com/order/catalog/product/A5322301). Efficient-Pro AGT Feed 1 has been demonstrated to improve yield and other end-points when used as described in the user guide (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/MAN0025907_EfficientProAGTFeed1_UG.pdf).
For research use only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.