Learn More
Gibco™ Efficient-Pro™ AGT™ Medium (+) Insulin
Description
Efficient-Pro AGT Medium (+) Insulin is a dry-format next-generation Chinese Hamster Ovary (CHO) medium used in the production of proteins such as monoclonal antibodies (mAbs), offering significant performance and ease-of-use improvements compared to other products.
Designed to work optimally with the Efficient-Pro Feed system, Efficient-Pro AGT Medium (+) Insulin has been formulated specifically for use with insulin-dependent CHO cell lines. The Efficient-Pro system increases titer and specific productivity, improves protein quality, and is easier to use and scale with convenient formats and multiple packaging configurations available.
Features of Efficient-Pro AGT Medium (+) Insulin include:
- Benefit from the ease-of-use associated with Gibco Advanced Granulation technology (AGT) for auto-pH and simplified dissolution
- Supports the growth and productivity of insulin-dependent CHO cell lines
- Sustains cell viability for longer bioreactor runs and maximized titer
- Helps speed up process development and streamline transfer to manufacturing scale
- Increases flexibility with a range of format and packaging configurations
Efficient-Pro AGT Medium (+) Insulin is an animal-origin-free (AOF) cell culture medium based on the Efficient-Pro AGT Medium formulation with the addition of recombinant human insulin. It is designed for the bioproduction of mAbs and other therapeutic proteins in both batch and fed-batch bioprocessing workflows. It helps support the growth and productivity of insulin-dependent CHO cell lines by eliminating the need for manual insulin supplementation. This modification reduces handling steps, minimizes variability, and improves workflow efficiency. Designed to be paired with the Efficient-Pro Feed system, Efficient-Pro AGT Medium (+) Insulin provides the power to help achieve high titers, specific productivity, and the simplicity to start process development faster, facilitate easier adaptation, and transfer to manufacturing scale with ease.
Specifications
Specifications
| Cell Line | Insulin-dependent CHO Cell Lines |
| Classification | Chemically-defined, Animal Origin-free, Serum-free, cGMP |
| Form | Powder (AGT) |
| Product Type | CHO Media |
| Serum Level | Serum-free |
| Product Line | Next Gen CHO Media |
| Quantity | 100 L |
| Shipping Condition | Dry Ice |
| Green Features | Sustainable packaging |
| Content And Storage | Store in the dark at 2°C to 8°C. |
| Show More |
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.