Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Efficient-Pro™ AGT™ Medium

Catalog No. A5322304
Change view
Click to view available options
Quantity:
1 L
10 L
100 L
500 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A5322304 500 L
A5322301 1 L
A5322302 10 L
A5322303 100 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. A5322304 Supplier Gibco™ Supplier No. A5322304
Add to Cart
Edge
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Gibco™ Efficient-Pro™ Media delivers superior product performance, improved product titer, better protein quality and lot-to-lot consistency for more control and predictability. Chemically defined and completely scalable, available in Gibco™ AGT™ formats for sustained cell viability.

  • A next generation CHO media used in the production of monoclonal antibodies
  • AGT Medium offers significant performance and ease-of-use improvements not seen in existing Gibco™ or competitor products
  • Proprietary AGT™ format is the only one of its kind in the industry and is the most advanced media format available in the market
  • Formulated as a chemically-defined, animal-origin free cell culture media used in the bioproduction of mAbs & other therapeutics in traditional batch & fed-batch bioprocessing workflows
  • Free of growth factors and insulin to ensure batch-to-batch consistency and production efficiency
  • Designed to work optimally with Efficient-Pro Feed 1 and Efficient-Pro Feed 2 for use in CHO-K1 and DG-44/CHO-S cell lines respectively
  • The Efficient-Pro system increases titer and specific productivity, improves protein quality and metabolite profiles
  • System is easier to use with less solubility issues than other CHO media and feed offerings
  • Increase ease-of-use with Gibco’s™ trademarked, patented Advanced Granulation Technology™ (AGT™) for auto-pH and simplified dissolution
  • Sustain cell viability for longer bioreactor runs & maximized titer
  • Demonstrated to improve titers in a CHO-K1 cell line up to 147% when paired with Efficient-Pro™ Feed 1 compared to an industry-leader’s media and feed system
  • Improve downstream outputs such as protein glycosylation, charge-variants & aggregation
  • Speed-up process development and streamline transfer to manufacturing scale
  • Increase flexibility with a range of format & packaging configurations
  • Decrease cell shock with simplified adaptation protocols

Specifications

Cell Line CHO-DG44, CHO-S, CHO-K1, GS-CHO
Classification Animal Origin-free, Chemically-defined, Protein-free, Serum-free
Form Powder (AGT)
Quantity 500 L
Shipping Condition Room temperature
Content And Storage Store in the dark at 2–8°C.

Culture Type Mammalian Cell Culture
How should I reconstitute Efficient-Pro AGT Medium (Cat. No. A5322301, A5322302, A5322303, A5322304)?

Please follow the steps shown below to reconstitute Efficient-Pro AGT Medium (Cat. No. A5322301, A5322302, A5322303, A5322304) and to make Complete Medium:

- Measure 90% of the final volume of deionized or distilled water at room temperature (15-30 degrees C).
- Add Efficient-Pro AGT Medium at 27.3 grams/L to water.
- Mix for a minimum of 20 min.
- Use a calibrated vessel to dilute to final production volume with ambient deionized or distilled water.
- Mix for an additional 20 min.
- Measure the pH, then measure and record osmolality.
- Sterilize the product immediately by membrane filtration(positive pressure recommended).
Note: Once the product is filtered, use immediately or store at 2-8 degrees C for up to 6 months. Protect from light.

To make Complete Medium,
- Supplement Efficient-Pro AGT Medium with L‑glutamine or GlutaMAX Supplement at 2-8 mM final concentration prior to use (except for CHO-K1 GS cell lines).
Note: The recommended L‑glutamine and/or GlutaMAX Supplement concentration is 6 mM.
- Add 10 mL/L of HT Supplement for use in applications not requiring DHFR amplification.
- Glucose supplementation may be required for terminal batch cultures and should be determined empirically.
- Add Anti-Clumping Agent (1 mL/L) to the medium to reduce cell aggregation, if required.
Note: Consider reducing L‑glutamine concentration for fed-batch or perfusion protocols, or if the cell line in-use is sensitive to ammonia. Addition of a surfactant such as Pluronic™ F-68 is not required.

What is the shelf-life of Efficient-Pro AGT Medium?

The shelf-life of Efficient-Pro AGT Medium is 12 months from the date of manufacture. The expiration date is provided on the product label.

If my cell line is sensitive to ammonia, can I still use Efficient-Pro AGT Medium?

Yes, however, we recommend reducing the L-glutamine concentration if a decrease in cell viability is observed. The recommended glutamine concentration is 2-8 mM. Another way to reduced ammonia production is to use GlutaMAX Supplement instead of L-glutamine. Addition of Pluronic F-68 is not required.

Does Efficient-Pro AGT Medium require supplementation for use?

When using a CHO cell line with a selection system other than the glutamine synthetase (GS) system, we recommend adding 2-8 mM L-glutamine or GlutaMAX Supplement to Efficient-Pro AGT Medium. Additionally, we recommend making sure that glucose levels do not fall below 2 g/L in culture by supplementing with glucose as needed. Also, if clumping becomes an issue, then add anti-clumping agent (ACA) to your cultures at 1% v/v.

What is the concentration of glucose in Efficient Pro-AGT Medium?

Efficient Pro-AGT Medium contains 4.7 g/L of glucose.

Can CHO cells that are created or frozen in other production media be thawed into Efficient-Pro AGT Medium or adapted for growth and production in Efficient-Pro AGT Medium?

Cells that are grown in or adapted to, and frozen in other cell culture media readily adapt to Efficient-Pro AGT Medium. However, if your cells are experiencing a significant lag or delay in growth, please directly adapt the cells into Efficient-Pro AGT Medium and passage them for a minimum of 3 passages, until a consistent doubling time is observed.


For research use only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.