Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Efficient-Pro™ Medium (+) Insulin

Catalog No. A4000458502
Change view
Click to view available options
Quantity:
1000 mL
10 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A4000458502 10 L
A4000458501 1000 mL
A4000458503 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A4000458502 Supplier Gibco™ Supplier No. A4000458502
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Efficient-Pro Medium (+) Insulin is a next-generation Chinese Hamster Ovary (CHO) medium used in the production of proteins such as monoclonal antibodies (mAbs), offering significant performance and ease-of-use improvements compared to other products.

Efficient-Pro Medium (+) Insulin is a next-generation Chinese Hamster Ovary (CHO) medium used in the production of proteins such as monoclonal antibodies (mAbs), offering significant performance and ease-of-use improvements compared to other products.

Designed to work optimally with the Efficient-Pro Feed system, Efficient-Pro Medium (+) Insulin has been formulated specifically for use with insulin-dependent CHO cell lines. The Efficient-Pro system increases titer and specific productivity, improves protein quality, and is easier to use and scale with convenient formats and multiple packaging configurations available.

Features of Efficient-Pro Medium (+) Insulin include:

  • Supports the growth and productivity of insulin-dependent CHO cell lines
  • Sustains cell viability for longer bioreactor runs and maximized titer
  • Helps speed up process development and streamline transfer to manufacturing scale
  • Increases flexibility with a range of format and packaging configurations

Efficient-Pro Medium (+) Insulin is a chemically defined and animal-origin-free (AOF) cell culture medium based on the Efficient-Pro Medium formulation with the addition of recombinant human insulin. It is designed for the bioproduction of mAbs and other therapeutic proteins in both batch and fed-batch bioprocessing workflows. It helps support the growth and productivity of insulin-dependent CHO cell lines by eliminating the need for manual insulin supplementation. This modification reduces handling steps, minimizes variability, and helps improve workflow efficiency. Designed to be paired with the Efficient-Pro Feed system, Efficient-Pro Medium (+) Insulin provides the power to help achieve high titers, specific productivity, and the simplicity to start process development faster, facilitate easier adaptation, and transfer to manufacturing scale with ease.

Specifications

Cell Line Insulin-dependent CHO Cell Lines
Classification Chemically-defined, Animal Origin-free, Serum-free, cGMP
Form Liquid
Product Type CHO Media
Serum Level Serum-free
Product Line Next Gen CHO Media
Quantity 10 L
Shipping Condition Room temperature
Content And Storage Store in the dark at 2°C to 8°C.
Culture Type Mammalian Cell Culture

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.