Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Efficient-Pro™ Medium

Catalog No. A5322203
Change view
Click to view available options
Quantity:
1000 mL
10 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A5322203 20 L
A5322201 1000 mL
A5322202 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A5322203 Supplier Gibco™ Supplier No. A5322203
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Gibco™ Efficient-Pro™ Media delivers superior product performance, improved product titer, better protein quality and lot-to-lot consistency for more control and predictability. Chemically defined and completely scalable, available in liquid or AGT™ formats for sustained cell viability.

  • A next generation CHO media used in the production of monoclonal antibodies
  • Liquid medium offers significant performance and ease-of-use improvements not seen in existing Gibco™ or competitor products
  • Formulated as a chemically-defined, animal-origin free cell culture media used in the bioproduction of mAbs & other therapeutics in traditional batch & fed-batch bioprocessing workflows
  • Free of growth factors and insulin to ensure batch-to-batch consistency and production efficiency
  • Designed to work optimally with Efficient-Pro Feed 1 and Efficient-Pro Feed 2 for use in CHO-K1 and DG-44/CHO-S cell lines respectively
  • The Efficient-Pro system increases titer and specific productivity, improves protein quality and metabolite profiles
  • System is easier to use with less solubility issues than other CHO media and feed offerings
  • Sustain cell viability for longer bioreactor runs & maximized titer
  • Demonstrated to improve titers in a CHO-K1 cell line up to 147% when paired with Efficient-Pro™ Feed 1 compared to an industry-leader’s media and feed system
  • Improve downstream outputs such as protein glycosylation, charge-variants & aggregation
  • Speed-up process development and streamline transfer to manufacturing scale
  • Increase flexibility with a range of format & packaging configurations
  • Decrease cell shock with simplified adaptation protocols

Specifications

Cell Line CHO-DG44, CHO-S, CHO-K1, GS-CHO
Classification Animal Origin-free, Chemically-defined, Protein-free, Serum-free
Form Liquid
Quantity 20 L
Shipping Condition Room temperature
Content And Storage Store in the dark at 2–8°C.

Culture Type Mammalian Cell Culture
What is the shelf-life of Efficient-Pro Medium?

The shelf-life of Efficient-Pro Medium is 12 months from the date of manufacture. The expiration date is provided on the product label.

If my cell line is sensitive to ammonia, can I still use Efficient-Pro Medium?

Yes, however, we recommend reducing the L-glutamine concentration if a decrease in cell viability is observed. The recommended glutamine concentration is 2-8 mM. Another way to reduced ammonia production is to use GlutaMAX Supplement instead of L-glutamine. Addition of Pluronic F-68 is not required.

Does Efficient-Pro Medium require supplementation for use?

When using a CHO cell line with a selection system other than the glutamine synthetase (GS) system, we recommend adding 2-8 mM L-glutamine or GlutaMAX Supplement to Efficient-Pro Medium. Additionally, we recommend making sure that glucose levels do not fall below 2 g/L in culture by supplementing with glucose as needed. Also, if clumping becomes an issue, then add anti-clumping agent (ACA) to your cultures at 1% v/v.

What is the concentration of glucose in Efficient-Pro Medium?

Efficient-Pro Medium contains 4.7 g/L of glucose.

Can CHO cells that are created or frozen in other production media be thawed into Efficient-Pro Medium or adapted for growth and production in Efficient-Pro Medium?

Cells that are grown in or adapted to, and frozen in other cell culture media readily adapt to Efficient-Pro Medium. However, if your cells are experiencing a significant lag or delay in growth, please directly adapt the cells into Efficient-Pro Medium and passage them for a minimum of 3 passages, until a consistent doubling time is observed.

For research use only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.