Learn More
Med Vet International Effitix Topical Solution for Dogs, 45-88.9 lbs, 3 Month
Catalog No. 502188397
Encompass_Preferred
Effitix Topical Solution for Dogs contains Fipronil and Permethrin, which control infestatons caused by fleas, ticks and lice; repels biting flies and repels and kills mosquitoes.Products are not always manufacturer specific. Actual product appearance may differ from website image.
Catalog No. 50-218-8397 Supplier Med Vet International Supplier No. PHEFFITIXPINK
Effitix Topical Solution for Dogs contains Fipronil and Permethrin, which control infestatons caused by fleas, ticks and lice; repels biting flies and repels and kills mosquitoes.Products are not always manufacturer specific. Actual product appearance may differ from website image.