Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Effusanin A | 30220-43-0 | 1 MG
SDP

Catalog No. 5000420966
Encompass_Preferred

Effusanin A is a natural product isolated from *Isodon rugosus*. It exhibits DNA-damaging and antibacterial activities, intended for research use only. This product has a purity of 99.0%.

  • Purity of 99.0%
  • Natural product isolated from *Isodon rugosus*
  • Exhibits DNA-damaging properties
  • Exhibits antibacterial properties
  • Molecular weight: 348.43
  • Chemical formula: C20H28O5
  • Appearance: White to off-white solid
  • Classified as terpenoids, diterpenoids
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: 4°C, protect from light
  • In solvent: -80°C for 6 months or -20°C for 1 month (protect from light)

Catalog No. 50-004-20966 Supplier Medchemexpress LLC Supplier No. HYN37981MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Effusanin A is a natural product isolated from *Isodon rugosus*. It exhibits DNA-damaging and antibacterial activities, intended for research use only. This product has a purity of 99.0%.

  • Purity of 99.0%
  • Natural product isolated from *Isodon rugosus*
  • Exhibits DNA-damaging properties
  • Exhibits antibacterial properties
  • Molecular weight: 348.43
  • Chemical formula: C20H28O5
  • Appearance: White to off-white solid
  • Classified as terpenoids, diterpenoids
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: 4°C, protect from light
  • In solvent: -80°C for 6 months or -20°C for 1 month (protect from light)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.