Learn More
Medchemexpress LLC Efgartigimod alfa | 1821402-21-4 | 99.5% | 10MG
SDP
Catalog No. 5000259528
Encompass_Preferred
Efgartigimod alfa is an anti-neonatal Fc receptor (FcRn) IgG1 Fc fragment supplied as a research reagent for studying FcRn-mediated IgG recycling and autoimmune conditions such as myasthenia gravis. The material is provided as a purified recombinant protein with analytical characterization for in vitro and preclinical research use.
  • High purity (99.47% by SEC-HPLC) for reproducible results.
  • Characterized molecular weight (51.28 kDa) to support analytical assays.
  • Validated for FcRn/Fc receptor research applications.
  • Supplied in small-scale quantities suitable for laboratory studies.
  • Accompanied by product datasheet and batch-specific COA.
Catalog No. 50-002-59528 Supplier Medchemexpress LLC Supplier No. HY14730510MG
Efgartigimod alfa is an anti-neonatal Fc receptor (FcRn) IgG1 Fc fragment supplied as a research reagent for studying FcRn-mediated IgG recycling and autoimmune conditions such as myasthenia gravis. The material is provided as a purified recombinant protein with analytical characterization for in vitro and preclinical research use.
  • High purity (99.47% by SEC-HPLC) for reproducible results.
  • Characterized molecular weight (51.28 kDa) to support analytical assays.
  • Validated for FcRn/Fc receptor research applications.
  • Supplied in small-scale quantities suitable for laboratory studies.
  • Accompanied by product datasheet and batch-specific COA.