Learn More
Aviva Systems Biology Corp EFHA2 Antibody - N-terminal region (ARP55801_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55801_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EFHA2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human EFHA2. Alternate names: DKFZp313A0139, EFHA2ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Pig, Rabbit, Yeast
Catalog No. 50-014-61618