Learn More
Aviva Systems Biology Corp EFHC1 Antibody - N-terminal region (ARP53728_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53728_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EFHC1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human EFHC1. Alternate names: FLJ10466, FLJ37290, dJ304B14.2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-874