Learn More
Aviva Systems Biology Corp EFHD2 Antibody - N-terminal region (ARP58617_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58617_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against EFHD2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human EFHD2. Alternate names: MGC4342, RP3-467K16.3, SWS1ApplicationWB. Species reactivity: Cow, Human, Mouse, Pig, Rat, Yeast, Zebrafish
Catalog No. 50-014-62375