Learn More
Cayman Chemical Efinaconazole, Purity: ≥98%, CAS: 164650-44-6, 10 mg
SDP
Catalog No. 502994847
Encompass_Preferred
A triazole antifungal agent; inhibits the growth of T. rubrum and T. mentagrophytes clinical isolates (MICs of ≤2 to 60 ng/ml) and of C. albicans clinical isolates (MICs of ≤0.5 to >250 ng/ml); inhibits sterol 14α-demethylase and arrests ergosterol biosynthesis at the fungal membrane; inhibits ergosterol biosynthesis in T. mentagrophytes and C. albicans with (IC50s = 7.0 and 0.40 ng/ml, respectfully)
Catalog No. 50-299-4847 Supplier Cayman Chemical Supplier No. 2383910 MG
A triazole antifungal agent; inhibits the growth of T. rubrum and T. mentagrophytes clinical isolates (MICs of ≤2 to 60 ng/ml) and of C. albicans clinical isolates (MICs of ≤0.5 to >250 ng/ml); inhibits sterol 14α-demethylase and arrests ergosterol biosynthesis at the fungal membrane; inhibits ergosterol biosynthesis in T. mentagrophytes and C. albicans with (IC50s = 7.0 and 0.40 ng/ml, respectfully)