Learn More
Cayman Chemical Efinaconazole-d4, Purity: ≥99% deuterated forms (d1-d4), 1 mg
SDP
Catalog No. 502948835
Encompass_Preferred
An internal standard for the quantification of efinaconazole by GC- or LC-MS
Catalog No. 50-294-8835 Supplier Cayman Chemical Supplier No. 336381 MG
An internal standard for the quantification of efinaconazole by GC- or LC-MS