Learn More
Medchemexpress LLC HY-P7381 10ug Medchemexpress, Kallikrein-11 (Sf9 insect cells-expressed), Human CAS: Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP738110UG
Encompass_Preferred
Medchemexpress, HY-B0744B 100mg Eflornithine (HCl, hydrate) CAS:96020-91-6 Eflornithine hydrochloride, hydrate is a specific, irreversible inhibitor of the enzyme ornithine decarboxylase. Eflornithine is a medication for the treatment of African trypanosomiasis and excessive facial hair growth in women. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0605