Learn More
Medchemexpress LLC HY-B0744B 500mg , Eflornithine (hydrochloride hydrate) CAS:96020-91-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0744B500MG
Encompass_Preferred
HY-B0744B 500mg Eflornithine (hydrochloride hydrate) CAS: 96020-91-6 Eflornithine hydrochloride hydrate (DFMO hydrochloride hydrate) is a specific, irreversible inhibitor of the enzyme ornithine decarboxylase. Eflornithine hydrochloride hydrate is a medication for the treatment of African trypanosomiasis and excessive facial hair growth in women. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1144